Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kynu
Entrez Gene ID
116682
Gene Name
kynureninase
Gene Aliases
-
Chromosome Location
Chr.3: 28416926 - 28566939 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053902.2 NP_446354.1 5 - 6 486 133
XM_006234138.2 XP_006234200.1 5 - 6 549 133
XM_008761695.1 XP_008759917.1 5 - 6 549 133
GenBank mRNA BC078762.1 - 5 - 6 469 133
U68168.1 - 5 - 6 487 133

View Full Product Details