The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135761.1 | NP_001129233.1 | 10 - 11 | 3384 | 137 |
XM_006234103.2 | XP_006234165.1 | 9 - 10 | 3515 | 137 |