Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cops4
Entrez Gene ID
360915
Gene Name
COP9 signalosome subunit 4
Gene Aliases
-
Chromosome Location
Chr.14: 10718863 - 10749123 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004275.1 NP_001004275.1 8 - 9 1042 82
XM_006250667.2 XP_006250729.1 7 - 8 952 82
GenBank mRNA BC079384.1 - 8 - 9 1042 82

View Full Product Details