The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012946.1 | NP_037078.1 | 10 - 11 | 2222 | 132 |
GenBank mRNA | BC061755.1 | - | 10 - 11 | 2223 | 132 |
U27562.1 | - | 10 - 11 | 2222 | 132 |