Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc22a6
Entrez Gene ID
29509
Gene Name
solute carrier family 22 (organic anion transporter), member 6
Gene Aliases
Oat1, Orctl1, Paht, Roat1
Chromosome Location
Chr.1: 224824809 - 224833284 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017224.2 NP_058920.1 4 - 5 834 70
XM_006230978.2 XP_006231040.1 4 - 5 1089 70
GenBank mRNA AB004559.1 - 4 - 5 1064 70
AF008221.1 - 4 - 5 1042 70
BC104692.1 - 4 - 5 834 70

View Full Product Details