Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc22a6
Entrez Gene ID
29509
Gene Name
solute carrier family 22 (organic anion transporter), member 6
Gene Aliases
Oat1, Orctl1, Paht, Roat1
Chromosome Location
Chr.1: 224824809 - 224833284 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017224.2 NP_058920.1 9 - 10 1611 88
XM_006230978.2 XP_006231040.1 9 - 10 1734 88
GenBank mRNA AB004559.1 - 9 - 10 1841 88
AF008221.1 - 9 - 10 1819 88
BC104692.1 - 9 - 10 1611 88

View Full Product Details