Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfpl1
Entrez Gene ID
684755
Gene Name
zinc finger protein-like 1
Gene Aliases
-
Chromosome Location
Chr.1: 221444588 - 221448548 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286936.1 NP_001273865.1 2 - 3 213 75
XM_006230901.2 XP_006230963.1 2 - 3 189 75
XM_008760175.1 XP_008758397.1 2 - 3 177 75
GenBank mRNA CB717504.1 - 2 - 3 213 75
FQ227385.1 - 2 - 3 249 75

View Full Product Details