Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Aoc1
- Entrez Gene ID
-
65029
- Gene Name
- amine oxidase, copper containing 1
- Gene Aliases
-
Abp, Abp1
- Chromosome Location
-
Chr.4: 78496043 - 78515585 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X73912.1 |
- |
1 - 2 |
48 |
67 |
View Full Product Details