Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpini1
Entrez Gene ID
116459
Gene Name
serpin peptidase inhibitor, clade I (neuroserpin), member 1
Gene Aliases
PI-12, raPIT5a
Chromosome Location
Chr.2: 174013378 - 174111755 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053779.1 NP_446231.1 2 - 3 363 83
XM_008761076.1 XP_008759298.1 3 - 4 650 83
GenBank mRNA AF193014.1 - 2 - 3 363 83
AF193015.1 - 2 - 3 369 83
BC061536.1 - 2 - 3 377 83

View Full Product Details