Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nab1
Entrez Gene ID
64824
Gene Name
Ngfi-A binding protein 1
Gene Aliases
-
Chromosome Location
Chr.9: 53906073 - 53945473 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006244902.2 XP_006244964.1 5 - 6 1378 63
GenBank mRNA FQ222344.1 - 6 - 7 1354 63

View Full Product Details