Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpgs1
Entrez Gene ID
691093
Gene Name
tubulin polyglutamylase complex subunit 1
Gene Aliases
Gtrgeo22, PGs1
Chromosome Location
Chr.7: 12912251 - 12918173 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109621.1 NP_001103091.1 1 - 2 814 115

View Full Product Details