Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grin3b
Entrez Gene ID
170796
Gene Name
glutamate receptor, ionotropic, N-methyl-D-aspartate 3B
Gene Aliases
GluN3B, NMDAR3B, NR3B
Chromosome Location
Chr.7: 12764993 - 12771310 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133308.2 NP_579842.2 8 - 9 2777 141
GenBank mRNA AF440691.2 - 8 - 9 2777 141

View Full Product Details