Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkx3-2
Entrez Gene ID
682329
Gene Name
NK3 homeobox 2
Gene Aliases
-
Chromosome Location
Chr.14: 73813531 - 73817888 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770239.1 XP_008768461.1 1 - 2 437 62

View Full Product Details