Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Drgx
Entrez Gene ID
252880
Gene Name
dorsal root ganglia homeobox
Gene Aliases
Drg11, Prrxl1
Chromosome Location
Chr.16: 8823872 - 8853430 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145767.1 NP_665710.1 6 - 7 811 56
GenBank mRNA U29174.1 - 6 - 7 811 56

View Full Product Details