Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zp1
Entrez Gene ID
85271
Gene Name
zona pellucida glycoprotein 1 (sperm receptor)
Gene Aliases
-
Chromosome Location
Chr.1: 226966366 - 226972746 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053509.1 NP_445961.1 1 - 2 205 109
GenBank mRNA AB000928.1 - 1 - 2 205 109

View Full Product Details