Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd22
Entrez Gene ID
294093
Gene Name
ankyrin repeat domain 22
Gene Aliases
-
Chromosome Location
Chr.1: 252430750 - 252461346 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191638.1 NP_001178567.1 2 - 3 349 132

View Full Product Details