Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Orm1
Entrez Gene ID
24614
Gene Name
orosomucoid 1
Gene Aliases
AGP, Agpa1, OMD, Orm
Chromosome Location
Chr.5: 79179668 - 79182820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053288.2 NP_445740.1 4 - 5 466 102
GenBank mRNA FQ209739.1 - 4 - 5 468 102
FQ210338.1 - 4 - 5 102
FQ218200.1 - 4 - 5 471 102
FQ218302.1 - 4 - 5 468 102
FQ219945.1 - 4 - 5 472 102
FQ220896.1 - 4 - 5 472 102
FQ221011.1 - 4 - 5 471 102
FQ221095.1 - 4 - 5 467 102
FQ221803.1 - 3 - 4 262 102
FQ221848.1 - 4 - 5 468 102
FQ222215.1 - 4 - 5 468 102
FQ222231.1 - 3 - 4 279 102
FQ222829.1 - 4 - 5 468 102
FQ223035.1 - 4 - 5 468 102
FQ228353.1 - 4 - 5 382 102
FQ228371.1 - 4 - 5 472 102
FQ228464.1 - 4 - 5 466 102
FQ228466.1 - 4 - 5 468 102
FQ228489.1 - 4 - 5 468 102
FQ228584.1 - 4 - 5 468 102
FQ229026.1 - 4 - 5 467 102
FQ229053.1 - 3 - 4 192 102
FQ229515.1 - 4 - 5 386 102
FQ230009.1 - 4 - 5 472 102
J00696.1 - 4 - 5 467 102
V01216.1 - 4 - 5 466 102

View Full Product Details