Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgg
Entrez Gene ID
24367
Gene Name
fibrinogen gamma chain
Gene Aliases
-
Chromosome Location
Chr.2: 181987080 - 181994523 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012559.2 NP_036691.2 1 - 2 103 140
XM_006232525.1 XP_006232587.1 1 - 2 257 140
GenBank mRNA BC078893.1 - 1 - 2 103 140
FQ219408.1 - 1 - 2 123 140
FQ219511.1 - 1 - 2 125 140
FQ219541.1 - 1 - 2 125 140
FQ219604.1 - 1 - 2 124 140
J00734.1 - 1 - 2 88 140

View Full Product Details