The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012994.2 | NP_037126.1 | 2 - 3 | 499 | 85 |
GenBank mRNA | BC105770.1 | - | 2 - 3 | 499 | 85 |
L27867.1 | - | 1 - 2 | 405 | 85 |