Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tk1
Entrez Gene ID
24834
Gene Name
thymidine kinase 1, soluble
Gene Aliases
Tk
Chromosome Location
Chr.10: 106817556 - 106828884 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052800.1 NP_434687.1 4 - 5 360 93
GenBank mRNA AW919734.1 - 2 - 3 197 93
CV072953.1 - 4 - 5 359 93
FQ229684.1 - 4 - 5 400 93
FQ234584.1 - - 370 93
FQ234769.1 - - 370 93

View Full Product Details