Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sept8
Entrez Gene ID
303135
Gene Name
septin 8
Gene Aliases
-
Chromosome Location
Chr.10: 38877422 - 38906480 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107002.1 NP_001100472.1 3 - 4 467 112
XM_006246265.2 XP_006246327.1 3 - 4 679 112
XM_006246266.2 XP_006246328.1 3 - 4 480 112
XM_006246270.2 XP_006246332.1 3 - 4 480 112
XM_006246271.2 XP_006246333.1 3 - 4 480 112
GenBank mRNA BC158796.1 - 3 - 4 443 112

View Full Product Details