Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dok7
Entrez Gene ID
305448
Gene Name
docking protein 7
Gene Aliases
RGD1566416
Chromosome Location
Chr.14: 80926071 - 80959398 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130062.1 NP_001123534.1 1 - 2 58 84

View Full Product Details