Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc19
Entrez Gene ID
502525
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 19
Gene Aliases
RGD1560220
Chromosome Location
Chr.2: 120504115 - 120531782 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134640.1 NP_001128112.1 5 - 6 414 73

View Full Product Details