Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ech1
Entrez Gene ID
64526
Gene Name
enoyl CoA hydratase 1, peroxisomal
Gene Aliases
Pxel
Chromosome Location
Chr.1: 87009798 - 87015996 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022594.1 NP_072116.1 2 - 3 267 95
GenBank mRNA BC062226.1 - 2 - 3 273 95
FQ214121.1 - 2 - 3 269 95
FQ230435.1 - 2 - 3 280 95
U08976.1 - 2 - 3 267 95

View Full Product Details