Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptov1
Entrez Gene ID
292888
Gene Name
prostate tumor overexpressed 1
Gene Aliases
-
Chromosome Location
Chr.1: 100859346 - 100865975 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008304.1 NP_001008305.1 4 - 5 560 67
GenBank mRNA BC085836.1 - 4 - 5 560 67

View Full Product Details