Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alad
Entrez Gene ID
25374
Gene Name
aminolevulinate dehydratase
Gene Aliases
ALADR, aminolevulinatedelta-dehydratase
Chromosome Location
Chr.5: 78368867 - 78379206 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238234.2 XP_006238296.1 1 - 2 689 130

View Full Product Details