Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcaf13
Entrez Gene ID
362902
Gene Name
DDB1 and CUL4 associated factor 13
Gene Aliases
Gm83, Wdsof1
Chromosome Location
Chr.7: 78005232 - 78040217 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173558.1 NP_001167029.1 3 - 4 442 164
XM_006241605.2 XP_006241667.1 3 - 4 401 164
GenBank mRNA EV763930.1 - 3 - 4 442 164
FQ228874.1 - 3 - 4 452 164
FQ229670.1 - 1 - 2 101 164

View Full Product Details