The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130565.1 | NP_001124037.1 | 4 - 5 | 884 | 67 |
GenBank mRNA | FQ222515.1 | - | 3 - 4 | 500 | 67 |