Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mtx1
Entrez Gene ID
295241
Gene Name
Metaxin 1
Gene Aliases
-
Chromosome Location
Chr.2: 188522605 - 188528765 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100667.1 NP_001094137.1 6 - 7 1114 69
XM_006232612.2 XP_006232674.1 7 - 8 1833 69
XM_008761135.1 XP_008759357.1 6 - 7 1965 69
GenBank mRNA BC099090.1 - 7 - 8 1118 69
BC158632.1 - 6 - 7 1114 69
FQ216199.1 - 4 - 5 502 69
FQ216274.1 - 7 - 8 1098 69

View Full Product Details