Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc50a1
Entrez Gene ID
295245
Gene Name
solute carrier family 50 (sugar efflux transporter), member 1
Gene Aliases
RGD1308478, Rag1ap1
Chromosome Location
Chr.2: 188585118 - 188587498 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106445.1 NP_001099915.1 5 - 6 667 59

View Full Product Details