Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hapln2
Entrez Gene ID
64057
Gene Name
hyaluronan and proteoglycan link protein 2
Gene Aliases
Bral1
Chromosome Location
Chr.2: 187395104 - 187404813 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022285.1 NP_071621.1 1 - 2 83 100
XM_008761208.1 XP_008759430.1 2 - 3 206 100
GenBank mRNA AB049056.1 - 1 - 2 83 100

View Full Product Details