The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025685.1 | NP_001020856.1 | 5 - 6 | 1092 | 157 |
GenBank mRNA | BC098724.1 | - | 5 - 6 | 1092 | 157 |