Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4b2
Entrez Gene ID
450222
Gene Name
C-type lectin domain family 4, member B2
Gene Aliases
Aplra1
Chromosome Location
Chr.4: 156107720 - 156131226 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005896.1 NP_001005896.1 5 - 6 517 93
GenBank mRNA AY365132.1 - 5 - 6 517 93
AY494066.1 - 5 - 6 484 93
AY581050.1 - 5 - 6 484 93

View Full Product Details