Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ergic2
Entrez Gene ID
297728
Gene Name
ERGIC and golgi 2
Gene Aliases
RGD1310606
Chromosome Location
Chr.4: 182569086 - 182600258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024984.1 NP_001020155.1 10 - 11 993 153
GenBank mRNA BC083597.1 - 10 - 11 993 153
FQ213743.1 - 11 - 12 960 153
FQ227558.1 - 10 - 11 878 153
FQ229098.1 - 12 - 13 1075 153
FQ230170.1 - 12 - 13 1076 153

View Full Product Details