The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109352.1 | NP_001102822.1 | 10 - 11 | 2447 | 104 |
XM_006237311.2 | XP_006237373.1 | 11 - 12 | 2260 | 104 | |
GenBank mRNA | FQ212196.1 | - | 3 - 4 | 695 | 104 |