Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casc1
Entrez Gene ID
297720
Gene Name
cancer susceptibility candidate 1
Gene Aliases
Las1
Chromosome Location
Chr.4: 179440984 - 179476932 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106627.3 NP_001100097.2 6 - 7 478 87
XM_008763378.1 XP_008761600.1 2 - 3 915 87
GenBank mRNA BC160907.1 - 6 - 7 478 87
CV108923.1 - 6 - 7 477 87

View Full Product Details