Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec14l1
Entrez Gene ID
360668
Gene Name
SEC14-like 1 (S. cerevisiae)
Gene Aliases
RGD1563123
Chromosome Location
Chr.10: 106065712 - 106112857 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108309.1 NP_001101779.1 3 - 4 513 96
GenBank mRNA FQ212255.1 - 3 - 4 423 96

View Full Product Details