Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spop
Entrez Gene ID
287643
Gene Name
speckle-type POZ protein
Gene Aliases
-
Chromosome Location
Chr.10: 83231197 - 83311987 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100496.1 NP_001093966.1 8 - 9 999 140
XM_006247180.2 XP_006247242.1 9 - 10 1803 140
XM_006247181.2 XP_006247243.1 10 - 11 1110 140
XM_006247182.2 XP_006247244.1 9 - 10 1059 140
XM_008767968.1 XP_008766190.1 9 - 10 1155 140
XM_008767969.1 XP_008766191.1 10 - 11 1077 140
GenBank mRNA BC091435.1 - 6 - 7 725 140
BC166743.1 - 8 - 9 999 140
FQ217100.1 - 10 - 11 1307 140

View Full Product Details