The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108279.1 | NP_001101749.1 | 6 - 7 | 1182 | 149 |
XM_008767841.1 | XP_008766063.1 | 7 - 8 | 1277 | 149 | |
XM_008767842.1 | XP_008766064.1 | 7 - 8 | 1567 | 149 | |
XM_008767843.1 | XP_008766065.1 | 7 - 8 | 1267 | 149 | |
GenBank mRNA | FQ223808.1 | - | 7 - 8 | 1238 | 149 |
FQ224714.1 | - | 7 - 8 | 1238 | 149 |