Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gfap
Entrez Gene ID
24387
Gene Name
glial fibrillary acidic protein
Gene Aliases
-
Chromosome Location
Chr.10: 90990762 - 90999501 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017009.2 NP_058705.2 4 - 5 779 110
XM_008768261.1 XP_008766483.1 4 - 5 845 110
GenBank mRNA BC088851.1 - 4 - 5 779 110
EF094476.1 - 4 - 5 850 110
L27219.1 - 4 - 5 783 110
U03700.1 - 4 - 5 803 110

View Full Product Details