Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc35b1
Entrez Gene ID
287642
Gene Name
solute carrier family 35, member B1
Gene Aliases
UGTrel1
Chromosome Location
Chr.10: 83201532 - 83208605 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199081.1 NP_954512.1 4 - 5 452 112
GenBank mRNA AY349135.1 - 4 - 5 452 112
BC085347.1 - 4 - 5 487 112
D87991.1 - 4 - 5 445 112
FQ233464.1 - 4 - 5 475 112

View Full Product Details