Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srsf2
Entrez Gene ID
494445
Gene Name
serine/arginine-rich splicing factor 2
Gene Aliases
Sfrs2
Chromosome Location
Chr.10: 105792779 - 105795985 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ232539.1 - 4 - 5 1054 87

View Full Product Details