Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasgef1c
Entrez Gene ID
360519
Gene Name
RasGEF domain family, member 1C
Gene Aliases
-
Chromosome Location
Chr.10: 35392762 - 35459267 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108273.1 NP_001101743.1 4 - 5 562 55

View Full Product Details