The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001513.2 | NP_001001513.2 | 6 - 7 | 793 | 94 |
XM_006246745.2 | XP_006246807.1 | 6 - 7 | 1036 | 94 | |
GenBank mRNA | AY607588.1 | - | 6 - 7 | 500 | 94 |
BC079107.1 | - | 6 - 7 | 793 | 94 | |
FQ221626.1 | - | 6 - 7 | 422 | 94 |