Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbx21
Entrez Gene ID
303496
Gene Name
T-box 21
Gene Aliases
-
Chromosome Location
Chr.10: 85032799 - 85049331 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107043.1 NP_001100513.1 1 - 2 635 67

View Full Product Details