Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wap
Entrez Gene ID
114596
Gene Name
whey acidic protein
Gene Aliases
-
Chromosome Location
Chr.14: 86897948 - 86900412 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053751.4 NP_446203.1 1 - 2 99 99
XM_008770244.1 XP_008768466.1 1 - 2 466 99
GenBank mRNA J00802.1 - 1 - 2 99 99

View Full Product Details