Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ghsr
- Entrez Gene ID
-
84022
- Gene Name
- growth hormone secretagogue receptor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 113066885 - 113070261 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U94321.1 |
- |
1 - 1 |
2872 |
101 |
View Full Product Details