Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnmb3
Entrez Gene ID
310303
Gene Name
potassium channel subfamily M regulatory beta subunit 3
Gene Aliases
-
Chromosome Location
Chr.2: 118868225 - 118882562 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104560.2 NP_001098030.1 3 - 4 80
GenBank mRNA AB297662.1 - 3 - 4 80

View Full Product Details