Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spata16
Entrez Gene ID
294932
Gene Name
spermatogenesis associated 16
Gene Aliases
RGD1563726
Chromosome Location
Chr.2: 112281328 - 112659911 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760900.1 XP_008759122.1 10 - 11 1768 106
XM_008760901.1 XP_008759123.1 10 - 11 1731 106

View Full Product Details