Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmcc1
Entrez Gene ID
312654
Gene Name
transmembrane and coiled-coil domain family 1
Gene Aliases
RGD1311909
Chromosome Location
Chr.4: 147920987 - 148090352 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006237072.2 XP_006237134.1 1 - 2 568 63

View Full Product Details